Learn More
Sigma Aldrich Fine Chemicals Biosciences Melatonin, 73-31-4, MFCD00005655, 250 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M5250250MG
Encompass
Empirical Formula (Hill Notation): C13H16N2O2, Molecular Weight: 232.28, powder, ≥98% (TLC), mp: 116.5-118 °C, storage temp: −20°C, Synonym: N-Acetyl-5-methoxytryptamine.
Catalog No. NC1298581