Learn More
Cayman Chemical MelatonIn-d4 1mg
SDP
Supplier:  Cayman Chemical 222911 MG
Encompass_Preferred
An internal standard for the quantification of melatonin by GC- or LC-MS
Catalog No. 50-298-7864