Learn More
Medchemexpress LLC Meldonium dihydrate | 86426-17-7 | MFCD13461779 | 99.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1836A
Encompass_Preferred
Meldonium (MET-88) dihydrate functions as a cardioprotective agent by competitively inhibiting γ-butyrobetaine hydroxylase (BBOX) and carnitine/organic cation transporter type 2 (OCTN2). It exhibits IC50 values of 34-62 μM for human recombinant BBOX and an EC50 of 21 μM for human OCTN2. Meldonium dihydrate is a fatty acid oxidation inhibitor.
  • Functions as a cardioprotective agent
  • Competitively inhibits γ-butyrobetaine hydroxylase (BBOX)
  • Competitively inhibits carnitine/organic cation transporter type 2 (OCTN2)
  • Exhibits IC50 values of 34-62 μM for human recombinant BBOX
  • Shows an EC50 of 21 μM for human OCTN2
  • Identified as a fatty acid oxidation inhibitor
Catalog No. 50-004-14081