Learn More
Cayman Chemical Melengestrol Acetate 100mg
SDP
Supplier:  Cayman Chemical 35471100 MG
Encompass_Preferred
A synthetic progestogen; binds to progesterone receptors in rhesus monkey myometrial cytosolic preparations with high affinity relative to progesterone; suppresses estrus in rats at 0.1 mg/animal per day; reduces tumor growth in androgen-dependent and -independent prostate tumors in rats at 10 mg/kg
Catalog No. 50-290-9773