Learn More
Medchemexpress LLC Melengestrol acetate | 2919-66-6 | 99.8% | 396.52 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY111614100MG
Encompass_Preferred
Melengestrol acetate is a progesterone derivative that acts as an orally active corticosteroid hormone. It promotes endometrial proliferation, maintains pregnancy, and delays menstrual activity. It is used as a contraceptive agent for growth-promoting effects and estrus suppression in animals. Melengestrol acetate also inhibits both androgen-dependent and -independent prostatic tumors in vivo, making it useful for cancer research.
  • Progesterone derivative
  • Orally active corticosteroid hormone
  • Promotes endometrial proliferation
  • Maintains pregnancy
  • Delays menstrual activity
  • Contraceptive agent for growth-promoting effects and estrus suppression in animals
  • Inhibits androgen-dependent and -independent prostatic tumors in vivo
  • Useful for cancer research
Catalog No. 50-004-39284