Learn More
Selleck Chemical LLC Melevodopa hydrochloride E4010-25MG
SDP
Supplier:  Selleck Chemical LLC E401025MG
Encompass_Preferred
L-3 4-Dihydroxyphenylalanine methyl ester hydrochloride(Melevodopa hydrochloride) is a precursor to L-DOPA It is an antiparkinsonian agent L-DOPA methyl ester that elicits antitumor functionality in leukemia and melanoma
Catalog No. 50-000-42123