Learn More
Medchemexpress LLC Melevodopa hydrochloride | 1421-65-4 | 99.8% | 247.68 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01196310MM/1ML
Encompass_Preferred
Melevodopa hydrochloride is an effervescent Levodopa prodrug. Levodopa is an orally active metabolic precursor of neurotransmitters dopamine and has anti-allodynic effects and the potential for Parkinson's disease.
  • Effervescent levodopa prodrug
  • Orally active metabolic precursor of neurotransmitters dopamine
  • Has anti-allodynic effects
  • Potential for Parkinson's disease treatment
  • Affects neurological disease areas
  • Involves GPCR/G protein and neuronal signaling
  • Acts on dopamine receptors
Catalog No. 50-004-52232