Learn More
Medchemexpress LLC Melinamide | 14417-88-0 | 99.9% | 383.61 g·mol-1 | C26H41NO | 25 MG

Supplier: Medchemexpress LLC HY10172225MG
Melinamide is an amide derivative of an unsaturated long-chain fatty acid used as a research tool to inhibit cholesterol absorption. It shows an IC50 of 20.9 μM for cholesterol uptake and inhibits acyl CoA:cholesterol acyltransferase (ACAT) activity in mucosal microsomes. For laboratory research use only.
- Purity 99.9%.
- CAS number 14417-88-0.
- Chemical formula C26H41NO.
- Molecular weight 383.61 g·mol-1.
- Activity IC50 20.9 μM for cholesterol absorption inhibition.
- Additional ACAT inhibition reported (50% inhibition at ~0.5 μM).
- Available as small-scale mg quantities and as a DMSO solution for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.