Learn More
Medchemexpress LLC Melitracen hydrochloride | 10563-70-9 | MFCD00242905 | >98.0% | 327.89 g/mol | C21H26ClN | 5 MG

Supplier: Medchemexpress LLC HY1082565MG
Melitracen hydrochloride is the hydrochloride salt of melitracen, a tricyclic antidepressant provided as a research standard for pharmacological and biochemical studies. It is used to investigate monoamine uptake and receptor interactions and is supplied as a solid suitable for analytical, assay, and formulation research applications.
- High purity suitable for analytical use (>98% HPLC).
- Molecular weight 327.89 g/mol for stoichiometric calculations.
- Chemical formula C21H26ClN.
- Stable solid form for long-term storage.
- Suitable for pharmacology and receptor binding studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.