Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Melitracen hydrochloride | 10563-70-9 | MFCD00242905 | 99.8% | 327.89 g·mol⁻¹ | C21H26ClN | 500 MG
SDP

Supplier:  Medchemexpress LLC HY108256500MG

Encompass_Preferred

Melitracen hydrochloride is an orally active tricyclic antidepressant research compound that inhibits norepinephrine and serotonin reuptake. Supplied as the hydrochloride salt, it is intended for in vitro and preclinical investigation of monoaminergic activity.

  • Inhibits norepinephrine and serotonin reuptake, increasing monoamine availability.
  • Provided as the hydrochloride salt for improved stability and solubility.
  • High reported purity (≈99.8%) suitable for research applications.
  • Molecular formula C21H26ClN, molecular weight 327.89 g·mol⁻¹.
  • Soluble in DMSO and water at high concentrations with sonication.
  • Recommended sealed storage away from moisture; short-term storage in solution at low temperatures.

Catalog No. 50-002-42779


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.