Learn More
Selleck Chemical LLC Melitracen hydrochloride S5034-25mg
SDP
Supplier:  Selleck Chemical LLC S503425MG
Encompass_Preferred
Melitracen hydrochloride (Thymeol) is a potential dopamine D1/2 receptor antagonist which has antidepressant properties
Catalog No. 50-311-2048