Learn More
Cayman Chemical MelItracenhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 4011710 MG
Encompass_Preferred
A thymoleptic and tricyclic antidepressant; inhibits 5-HT uptake in rat brain synaptosomes (IC50 = 670 nM); increases the lethality of yohimbine in mice at 20 and 40 mg/kg
Catalog No. 50-294-2446