Learn More
Selleck Chemical LLC Melittin-5mg
SDP
Supplier:  Selleck Chemical LLC S97945MG
Encompass_Preferred
Melittin (MLT, Forapin, Forapine) is an activator of phospholipase A2 (PLA2) that stimulates the activity of the low molecular weight PLA2, while it does not the increase the activity of the high molecular weight enzyme.
Catalog No. 50-238-9962