Learn More
TARGETMOL CHEMICALS INC MELK-8A HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T119965MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MELK-8a hydrochloride is a potent inhibitor of maternal embryonic leucine zipper kinase (MELK IC50 = 2 nM). purity: 99%
Catalog No. 50-258-1492