Learn More
Medchemexpress LLC Meloside A (Isovitexin 2''-O-glucoside) | 60767-80-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN51245MG
Encompass_Preferred
Meloside A, also known as Isovitexin 2''-O-glucoside, is a phenylpropanoid found in barley known for its antioxidant activity. This compound can inhibit cell apoptosis and reduce reactive oxygen species (ROS) production. It also demonstrates an ability to inhibit androgen receptor (AR) nuclear translocation and AR protein expression. Furthermore, Meloside A can decrease the levels of IL-6, TGF-β1, and DKK-1, making it a valuable tool for research in inflammation and endocrinology, including studies related to hair loss.
  • Exhibits antioxidant activity
  • Inhibits cell apoptosis and ROS production
  • Inhibits androgen receptor (AR) nuclear translocation and expression
  • Reduces IL-6, TGF-β1, and DKK-1 levels
  • Useful for inflammation and endocrinology research
  • Does not induce significant cytotoxicity in HDPCs (5-100 μg/mL, 24 h)
Catalog No. 50-004-17784