Learn More
Medchemexpress LLC Meloxicam | 71125-38-7 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB026110MM/1ML
Encompass_Preferred
Meloxicam is a non-steroidal antiinflammatory agent that can pass through the blood-brain barrier. It functions by inhibiting COX activity, showing IC50s of 0.49 μM for COX-2 and 36.6 μM for COX-1. It is identified for laboratory use and suitable for the manufacture of substances.
  • Non-steroidal antiinflammatory agent
  • Passes through the blood-brain barrier
  • Inhibits COX activity
  • Targets COX, Autophagy, Apoptosis, MMP
  • Relevant in Immunology/Inflammation, Autophagy, Apoptosis, Metabolic Enzyme/Protease research
  • Identified for laboratory use
  • Suitable for substance manufacturing
Catalog No. 50-004-06892