Learn More
Medchemexpress LLC Meloxicam | 71125-38-7 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0261R50MG
Encompass_Preferred
Meloxicam (Standard) is the analytical standard of Meloxicam. This product is intended for research and analytical applications. Meloxicam is a non-steroidal antiinflammatory agent that inhibits COX activity, with IC50s of 0.49 μM and 36.6 μM for COX-2 and COX-1, respectively. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for Meloxicam
  • Intended for research and analytical applications
  • Inhibits COX activity
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-06153