Learn More
Cayman Chemical MeloxIcam-d3 500ug
SDP
Supplier:  Cayman Chemical 25508500 UG
Encompass_Preferred
An internal standard for the quantification of meloxicam by GC- or LC-MS
Catalog No. 50-301-5902