Learn More
Medchemexpress LLC Meloxicam (Standard) | 71125-38-7 | 99.89% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0261R100MG
Encompass_Preferred
Meloxicam (Standard) is the analytical standard of Meloxicam, intended for research and analytical applications. Meloxicam is a non-steroidal anti-inflammatory agent that inhibits COX activity, with IC50s of 0.49 μM for COX-2 and 36.6 μM for COX-1.
  • Intended for research and analytical applications
  • Non-steroidal anti-inflammatory agent
  • Inhibits COX activity
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-05380