Learn More
Med Vet International Meloxidyl (Meloxicam) Injection, 5mg/mL, 10mL, Each
Supplier:  Med Vet International RXMELOX10INJV
Encompass_Preferred
Meloxidyl (meloxicam) Injection 5 mg/mL is indicated for use in dogs for the control of pain and inflammation associated with osteoarthritis (OA).Meloxidyl (meloxicam) Injection 5 mg/mL is indicated for use in cats for the control of postoperative pain and inflammation associated with orthopedic surgery, ovariohysterectomy, and castration when administered prior to surgery.
Catalog No. 50-282-8314