Learn More
Cayman Chemical Melperon-d4hydrochlorIde 1mg
SDP
Supplier:  Cayman Chemical 314891 MG
Encompass_Preferred
An internal standard for the quantification of melperone by GC- or LC-MS
Catalog No. 50-299-1920