Learn More
Medchemexpress LLC Melperone-d4 (hydrochloride) | 1219798-80-7 | 99.8% | 500 UG
SDP
Supplier:  Medchemexpress LLC HYB2169S2500UG
Encompass_Preferred
Melperone-d4 hydrochloride is the deuterium labeled Melperone hydrochloride. Melperone hydrochloride is a butyrophenone with atypical antipsychotic properties. It is a multireceptor antagonist with Kds of 102 nM, 180 nM, 180 nM, and 150 nM for 5-HT2A, dopamine D2, α1-adrenergic, and α2-adrenergic receptors, respectively. Melperone hydrochloride has weak binding to histamine H1, 5-HT2C, 5-HT1A, 5-HT1D, and muscarinic receptors, with Kd values ​​of 580 nM, 2100 nM, 2200 nM, 3400 nM, >10000 nM, respectively. It is also a CYP2D6 inhibitor and can be used for the study of schizophrenia and agitation in the elderly.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-12337