Learn More
United States Biological Corporation MEM EAGLE D VAL MOD W LGLT 25L
Supplier:  United States Biological Corporation M38612025L
Encompass_Preferred
Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.� Early attempts to cultivate normal mammalian fibroblasts and certain subtypes of HeLa cells revealed that they had specific nutritional requirements that could not be met by Eagle's Basal Medium (BME).�Subsequent studies using these and other cells in culture indicated that additions to BME could be made to aid growth of a wider variety of fastidious cells.�MEM, which incorporates these modifications, includes higher concentrations of amino acids so that the medium more closely approximates the protein composition of mammalian cells. MEM has been used for cultivation of a wide variety of cells grown in monolayers.� Optional supplementation of non-essential amino acids to the formulations that incorporate either Hanks' or Eagles' salts has broadened the usefulness of this medium.
Catalog No. NC9930008