Learn More
Medchemexpress LLC Memantine-d6 (hydrochloride) | 1189713-18-5 | 99.8% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0365AS1MG
Encompass_Preferred
Memantine-d6 (hydrochloride) is a deuterium-labeled Memantine hydrochloride. It functions as a moderate affinity, uncompetitive NMDA receptor antagonist and inhibits CYP2B6 and CYP2D6. This compound is frequently used in research as a stable isotope-labeled standard and as a tracer for quantitative analysis.
  • Deuterium labeled
  • Moderate affinity, uncompetitive NMDA receptor antagonist
  • Inhibits CYP2B6 and CYP2D6
  • Stable isotope-labeled standard
  • Suitable as a tracer
  • Applicable for quantitative analysis (NMR, GC-MS, LC-MS)
Catalog No. 50-004-06286