Learn More
Selleck Chemical LLC Memantine HCl S2043-500mg
SDP
Supplier:  Selleck Chemical LLC S2043500MG
Encompass_Preferred
Memantine HCl is an antagonist of NMDAR It is also a CYP2B6 and CYP2D6 inhibitor for recombinant CYP2B6 and CYP2D6 with IC50 of 1 12 M and 242 4 M Ki of 0 51 M and 84 4 M respectively
Catalog No. 50-311-1102