Learn More
Revvity Health Sciences Inc HTRF Human IL-6 / IL6R Binding Kit, 500 Assay Points
Supplier:  Revvity Health Sciences Inc 64BDIL6PEG
Encompass
HTRF Human IL-6 / IL6R Binding Kit, 500 Assay Points
Catalog No. 50-281-2539