Learn More
Biotium MemBrite™ Fix 543/560 Cell Surface Staining Kit, 100 Reactions
Supplier:  Biotium 30094T
Encompass_Preferred
MemBrite Fix 543/560 is a orange fluorescent dye that covalently labels the surface of live cells. Unlike lipophilic membrane dyes, MemBrite is well-retained after fixation and permeabilization or methanol fixation. One vial for 100 labeling reactions (200 uL staining volume). Ex/Em 541/560 nm.
Catalog No. NC1541157