Learn More
Molecular Dimensions MEMGOLD
SDP
Supplier:  Molecular Dimensions MD141
Encompass
Protein Crystallography. MemGold screen HT-96 (96 x 1.1mL) is based on crystallization conditions data mined from the Protein Data Bank and contains 96 conditions covering a range of pH, PEGs and salt additives. Addresses the diversity of membrane proteins studied, both alpha- and beta - types.
Catalog No. NC9057977