Learn More
Memmert Memmert Humidity Chamber HCP105 - 105L - 115V, Stainless Steel Construction with Programmable TwinDISPLAY controller
SDP
Supplier:  Memmert HCP 105 115V
Encompass_Preferred
Memmert Humidity Chamber - HCP provides excellent control, making it easy to achieve uniform atmosphere and temperature distribution in simulating environmental condistions. This 107L (3.8 cu ft) model is the 115 volt, with working humidity ranges of 20 to 95% rh, within temperatures of at least 6¿C over ambient up to 90¿C with humidity control. Includes (2) Stainless Steel Perforated Shelf, Atmocontrol Software, Manual, Operation manual, Calibration Certificate, Power Cord, Water tank and connections!
Catalog No. 50-225-4398
Add to Cart
Request a Quote