Learn More
CPC Scientific H-Asp-Tyr-DTrp-Val-DTrp-DTrp-Lys-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP117A
Encompass
Potent and selective NK2 tachykinin receptor antagonist.SEQUENCE: H-Asp-Tyr-DTrp-Val-DTrp-DTrp-Lys-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: DY-DTrp-V-DTrp-DTrp-K-NH2MOLECULAR FORMULA: C57H68N12O10MOLECULAR WEIGHT: 1081.24STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [135306-85-3]SYNONYMS: MEN-10,376, (Tyr,D-Trp,Lys-NH)-Neurokinin A (4-10)RESEARCH AREA: NeuropeptidesREFERENCES:C.A. Maggi, J. Pharm. & Exp. Ther., 257, 1172 (1991)R. Monteau et al., European Journal of Pharmacology, 314, 41 (1996)SKU(s): NEUP-117A
Catalog No. 50-210-9949