Learn More
Medchemexpress LLC Menadione | 58-27-5 | 99.8% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB033210G
Encompass_Preferred
Menadione is a naphthoquinone that is converted into active vitamin K2 in the body. It is a potential anticancer agent and radiosensitizer.
  • Converts into active vitamin K2 in the body
  • Potential anticancer agent
  • Potential radiosensitizer
  • Human endogenous metabolite
  • Induces necrosis and apoptosis in AR4-2J cells
  • Inhibits cell proliferation in AR4-2J cells
  • Induces wild-type P53
Catalog No. 50-004-06378