Learn More
Medchemexpress LLC Menadione-d3 | 5172-16-7 | 99.1% | 1 MG
Supplier: Medchemexpress LLC HYB0332S1MG
Menadione-d3 is the deuterium labeled Menadione. Menadione, a synthetic naphthoquinone, can be converted to active vitamin K2 in vivo. It is for research use only and not sold to patients. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
- Can be used as a tracer.
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.