Learn More
Cayman Chemical MenaquInon 9 5mg
SDP
Supplier:  Cayman Chemical 182425 MG
Encompass_Preferred
A vitamin K2 analog; has been found in various bacteria; binds to soluble nitrate reductase; increases plasma prothrombin concentration in vitamin K-deficient rats
Catalog No. 50-296-7608