Learn More
Medchemexpress LLC Menaquinone-4-d7 | 1233937-25-1 | ≥99% | 500 UG
SDP
Supplier:  Medchemexpress LLC HYB2156S500UG
Encompass_Preferred
Menaquinone-4-d7 (Vitamin K2 (MK-4)-d7) is the deuterium labeled Menaquinone-4. Menaquinone-4 is a vitamin K, used as a hemostatic agent and as an adjunctive therapy for the pain of osteoporosis. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Menaquinone-4
  • Used as a hemostatic agent
  • Adjunctive therapy for pain of osteoporosis
  • Stable heavy isotopes used as tracers for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-12331