Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Menin protein, human (His) | >90.0% | 50UG
SDP

Supplier:  Medchemexpress LLC HYP70316450UG

Encompass_Preferred

Menin protein, human (His) is a recombinant human menin expressed in E. coli with an N-terminal His tag corresponding to residues M1-L615. Menin functions as a tumor suppressor and transcriptional regulator involved in histone H3K4 methylation and regulation of telomerase expression; this recombinant protein is provided as a filtered, stabilized solution for biochemical and molecular biology research.

  • Human sequence corresponding to M1-L615.
  • N-terminal His tag for purification and detection.
  • Expressed in E. coli for scalable production.
  • Purity >90% as determined by reducing SDS-PAGE.
  • Molecular weight ~68.9 kDa.
  • Supplied in stabilized formulation; store frozen to maintain activity.

Catalog No. 50-001-74619


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.