Learn More
TARGETMOL CHEMICALS INC MENIN-MLL INHIBITOR MI-2 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T264910MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Menin-MLL inhibitor MI-2 (MI2) is a potent menin-MLL interaction inhibitor with IC50 of 446 nM. purity: 97%
Catalog No. 50-258-0695