Learn More
ABclonal Technology Menin Rabbit pAb
SDP
Supplier:  ABclonal Technology A15290500UL
Encompass_Preferred
This gene encodes menin, a tumor suppressor associated with a syndrome known as multiple endocrine neoplasia type 1. Menin is a scaffold protein that functions in histone modification and epigenetic gene regulation. It is thought to regulate several pathways and processes by altering chromatin structure through the modification of histones.
Catalog No. 50-001-08180