Learn More
Selleck Chemical LLC (-)-Menthol S4714-100mg
SDP
Supplier:  Selleck Chemical LLC S4714100MG
Encompass_Preferred
(-)-Menthol is a levo isomer of menthol and used as a cooling agent that strongly activates TRPM8
Catalog No. 50-310-9205