Learn More
Medchemexpress LLC (-)-Menthone | 14073-97-3 | 50 G

Supplier: Medchemexpress LLC HYN791650G
(-)-Menthone is a monoterpene component found in the essential oil of maturing peppermint. It has been studied for its biological activity, including its effect on *Anopheles stephensi* larvae.
- Purity: 98.0%
- Molecular weight: 154.25
- Formula: C10H18O
- Appearance: Liquid, colorless to light yellow
- Solubility in DMSO: 100 mg/mL, requires ultrasonication
- Storage: Pure form at -20°C for 3 years; in solvent at -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.