Learn More
Aat Bioquest MeO-Succ-Arg-Pro-Tyr-AMC
SDP
Supplier:  Aat Bioquest 13460
Encompass_Preferred
MeO-Succ-Arg-Pro-Tyr-AMC is a sensitive fluorogenic substrate for chymotrypsin-like proteases.
Catalog No. 50-195-3457