Learn More
Cayman Chemical MeOSuc-AAPV-pNA 500mg
SDP
Supplier:  Cayman Chemical 17601500 MG
Encompass_Preferred
A highly sensitive peptide substrate for both human and mouse neutrophil elastase and PR3
Catalog No. 50-296-6278