Learn More
Cayman Chemical MeOSucAlaAlaProValAMC 10mg
SDP
Supplier:  Cayman Chemical 1490710 MG
Encompass_Preferred
A fluorogenic substrate for human leukocyte and porcine pancreatic elastase; excitation: 355-380 nm; emission 440-460 nm
Catalog No. 50-297-2980