Learn More
Medchemexpress LLC Mepazine hydrochloride | 2975-36-2 | >97.0% | 10 MG

Supplier: Medchemexpress LLC HY121282A250MG
Mepazine hydrochloride is a phenothiazine-derived small-molecule MALT1 protease inhibitor used in preclinical research to induce apoptosis and probe MALT1-dependent signaling. It is supplied as the monohydrochloride salt and intended for research use only.
- Cas number 2975-36-2.
- Chemical formula C19H23ClN2S.
- Molecular weight 346.92 g/mol.
- Reported MALT1 inhibitory activity (IC50 ≈0.83 μM for GST-MALT1 full length).
- Typical use: in vitro research reagent for mechanistic and cell-based studies.
- Available package sizes reported: 1 mg, 5 mg, 10 mg.
- Purity reported by suppliers: ≥97% (HPLC).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.