Learn More
Medchemexpress LLC Mepazine hydrochloride | 2975-36-2 | 99.3% | 100 MG

Supplier: Medchemexpress LLC HY121282A100MG
Mepazine hydrochloride is the hydrochloride salt of mepazine, described as a potent and selective MALT1 protease inhibitor for research use. Supplier documentation provides solubility, storage, and stock solution preparation guidance for laboratory studies.
- Hydrochloride salt; CAS 2975-36-2.
- Molecular formula C19H23ClN2S; molecular weight 346.92.
- Soluble in DMSO (50 mg/mL) and water with heating; PBS solubility reported.
- Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
- Datasheet provides preparation examples and lists 1 mg, 5 mg, and 10 mg quantities.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.