Learn More
Medchemexpress LLC Mepazine hydrochloride | 2975-36-2 | 99.3% | 25 MG

Supplier: Medchemexpress LLC HY121282A25MG
Mepazine hydrochloride is the hydrochloride salt of mepazine, a phenothiazine-derived small molecule reported to inhibit MALT1 protease and to reduce viability of ABC-DLBCL cells by enhancing apoptosis. It is supplied for research use only.
- Acts as a MALT1 protease inhibitor (reported IC50 0.83 μM and 0.42 μM against GST-MALT1 full length and 325-760, respectively).
- CAS number 2975-36-2.
- Molecular formula C19H23ClN2S.
- Molecular weight 346.92.
- Purity 99.25% (reported).
- Available in multiple milligram pack sizes including 25 mg.
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.