Learn More
Medchemexpress LLC Mepazine hydrochloride | 2975-36-2 | 99.3% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY121282A50MG
Encompass_Preferred
Mepazine hydrochloride is a small-molecule MALT1 protease inhibitor used in research to induce apoptosis and reduce viability in certain lymphoma cell models. It is supplied as the hydrochloride salt at high purity for in vitro studies.
  • Potent MALT1 protease inhibition with reported IC50 values for GST-MALT1 constructs.
  • High purity suitable for research applications.
  • Supplied as a hydrochloride salt to aid solubility in experimental buffers.
  • Molecular weight 346.92 and formula C19H23ClN2S.
  • Available in small pack sizes for laboratory testing and screening.
Catalog No. 50-002-34846