Learn More
Aviva Systems Biology Corp MEPE Antibody - middle region (ARP36796_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36796_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MEPE. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human MEPE. Alternate names: OF45ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-514