Learn More
Medchemexpress LLC (+)-Mephenytoin | 70989-04-7 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB1184A50MG
Encompass_Preferred
(S)-Mephenytoin is an anticonvulsive agent and a substrate of the cytochrome P450 (CYP) isoform CYP2C19, suitable for the analysis of cytochrome P450 metabolism. This product is provided as a white to off-white solid.
  • Anticonvulsive agent
  • Substrate of cytochrome P450 (CYP) isoform CYP2C19
  • Used for the analysis of cytochrome P450 metabolism
  • High purity (99.9%)
  • Available as a solid
Catalog No. 50-004-12833