Learn More
Medchemexpress LLC Mepiquat chloride | 24307-26-4 | MFCD00055543 | ≥97.0% | 149.66 g/mol | C7H16ClN | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01744850G
Encompass_Preferred
Mepiquat chloride (N,N-Dimethylpiperidinium chloride; PIX) is a quaternary ammonium plant growth regulator used to reduce stem elongation and overall plant height, helping to optimize canopy architecture and agronomic performance. It is provided as a purified chemical (CAS 24307-26-4) suitable for laboratory research and formulation studies.
  • Reduces plant height and internode elongation.
  • Improves canopy density and lodging resistance.
  • Suitable for agronomic research and formulation testing.
  • High purity (≥97.0%) for reliable experimental results.
  • Available as a research-scale quantity for laboratory use.
Catalog No. 50-002-99975