Learn More
Apexbio Technology LLC Mepiroxol 6968-72-5 5g
SDP
Supplier:  Apexbio Technology LLC A84685000
Encompass_Preferred
Mepiroxol is a small-molecule compound characterized by its ability to form specific interactions with target proteins It engages in salt bridge interactions with arginine residues (such as Arg-153 and Arg-74) and participates in hydrogen bonding via its hydroxyl group with protein residues like Asp-50 and Gly-96 supporting enhanced binding affinity Researchers utilize Mepiroxol for investigations into protein-ligand interaction mechanisms aiding structural biology analyses and guiding drug-design efforts
Catalog No. 50-000-95251